siRNA Details
| Gene Symbol: Epha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256432.1 | NP_001243361.1 | 3 | 918 | ||
| NM_177671.5 | NP_808339.2 | 3 | 918 | |||
| GenBank mRNA | AK039700.1 | - | 3 | 881 | ||
| AK143168.1 | - | 1803 | ||||
| BC116991.1 | - | 3 | 848 | 40126310 | ||
| BC119084.1 | - | 3 | 848 | 8733833 | ||

Epha10
Genomic Map