siRNA Details
| Gene Symbol: Ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177682.3 | NP_808350.1 | 2, 3 | 261 | ||
| GenBank mRNA | AK090008.1 | - | 2, 3 | 275 | ||
| AK146704.1 | - | 2, 3 | 294 | |||
| AK150793.1 | - | 2, 3 | 298 | |||
| AK152328.1 | - | 2, 3 | 297 | |||
| AK152576.1 | - | 2, 3 | 297 | |||
| AK153160.1 | - | 2, 3 | 298 | |||
| AK171671.1 | - | 1, 2 | 1895 | |||
| AK172392.1 | - | 1, 2 | 1894 | |||
| BC048077.1 | - | 2, 3 | 261 | 6308253 | ||

Ccz1
Genomic Map