siRNA Details
| Gene Symbol: Iqcj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177585.3 | NP_808253.1 | 4 | 766 | ||
| GenBank mRNA | AK052136.1 | - | 4 | 767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcj| Gene Symbol: Iqcj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177585.3 | NP_808253.1 | 4 | 766 | ||
| GenBank mRNA | AK052136.1 | - | 4 | 767 | ||