siRNA Details
| Gene Symbol: March11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035990.1 | - | 2 | 1098 | ||
| BC139422.1 | - | 2 | 1025 | 8862444 | ||
| BC139423.1 | - | 2 | 1025 | 8862445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March11| Gene Symbol: March11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035990.1 | - | 2 | 1098 | ||
| BC139422.1 | - | 2 | 1025 | 8862444 | ||
| BC139423.1 | - | 2 | 1025 | 8862445 | ||