siRNA Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177606.4 | NP_808274.2 | 30 | 4939 | ||
| GenBank mRNA | AK029252.1 | - | 30 | 4942 | ||
| AK086794.1 | - | 10 | 1770 | |||
| AK173337.1 | - | 27 | 4594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhh2| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177606.4 | NP_808274.2 | 30 | 4939 | ||
| GenBank mRNA | AK029252.1 | - | 30 | 4942 | ||
| AK086794.1 | - | 10 | 1770 | |||
| AK173337.1 | - | 27 | 4594 | |||