siRNA Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289738.1 | NP_001276667.1 | 11 | 1402 | ||
| NM_177656.4 | NP_808324.1 | 11 | 1399 | |||
| GenBank mRNA | AK078482.1 | - | 11 | 1399 | ||
| BC137826.1 | - | 10 | 1279 | 8860845 | ||
| BC144943.1 | - | 10 | 1317 | |||
| BC144944.1 | - | 10 | 1279 | 9053492 | ||
| BC144945.1 | - | 9 | 1213 | |||

6820408C15Rik
Genomic Map