siRNA Details
| Gene Symbol: CLNS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293.2 | NP_001284.1 | 7 | 1034 | ||
| NM_001311200.1 | NP_001298129.1 | 6 | 932 | |||
| NM_001311201.1 | NP_001298130.1 | 6 | 860 | |||
| NM_001311202.1 | NP_001298131.1 | 5 | 824 | |||
| GenBank mRNA | AF005422.1 | - | 7 | 972 | ||
| AL530926.3 | - | 6 | 878 | |||
| BC119634.1 | - | 7 | 991 | 40115653 | ||
| BC119635.1 | - | 7 | 991 | 40115655 | ||
| U17899.1 | - | 7 | 1016 | |||
| U53454.1 | - | 7 | 1026 | |||
| X91788.1 | - | 7 | 1030 | |||

CLNS1A
Genomic Map