siRNA Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177662.2 | NP_808330.1 | 8 | 2120 | ||
| GenBank mRNA | AK034490.1 | - | 8 | 2120 | ||
| AK049470.1 | - | 8 | 2092 | |||
| AK143683.1 | - | 9 | 2241 | |||
| AK166103.1 | - | 8 | 2089 | |||
| BC044664.1 | - | 8 | 2012 | |||
| BC098219.1 | - | 8 | 2094 | 1066724 | ||

Ctso
Genomic Map