siRNA Details
| Gene Symbol: Fam198a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199927.1 | NP_001186856.1 | 2 | 361 | ||
| NM_177743.5 | NP_808411.1 | 2 | 292 | |||
| GenBank mRNA | AK050223.1 | - | 2 | 280 | ||
| AK134876.1 | - | 2 | 347 | |||
| AK139038.1 | - | 2 | 362 | |||
| AK158406.1 | - | 2 | 294 | |||
| BC113767.1 | - | 1 | 141 | 40058404 | ||

Fam198a
Genomic Map