siRNA Details
| Gene Symbol: Gm5082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145878.1 | NP_001139350.1 | 3 | 1204 | ||
| NM_177808.4 | NP_808476.2 | 5 | 1367 | |||
| GenBank mRNA | AK037195.1 | - | 5 | 1367 | ||
| BC098182.1 | - | 3 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5082| Gene Symbol: Gm5082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145878.1 | NP_001139350.1 | 3 | 1204 | ||
| NM_177808.4 | NP_808476.2 | 5 | 1367 | |||
| GenBank mRNA | AK037195.1 | - | 5 | 1367 | ||
| BC098182.1 | - | 3 | 1204 | |||