siRNA Details
Gene Symbol: Zfp459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177811.4 | NP_808479.1 | 3 | 263 | ||
GenBank mRNA | AK036923.1 | - | 3 | 264 | ||
BC100389.1 | - | 3 | 229 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177811.4 | NP_808479.1 | 3 | 263 | ||
GenBank mRNA | AK036923.1 | - | 3 | 264 | ||
BC100389.1 | - | 3 | 229 |