siRNA Details
Gene Symbol: Gm4894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177701.3 | NP_808369.1 | 4 | 596 | ||
GenBank mRNA | AK039996.1 | - | 4 | 597 | ||
BC132633.1 | - | 4 | 544 | 40130910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177701.3 | NP_808369.1 | 4 | 596 | ||
GenBank mRNA | AK039996.1 | - | 4 | 597 | ||
BC132633.1 | - | 4 | 544 | 40130910 |