siRNA Details
Gene Symbol: Zfp874a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177712.4 | NP_808380.1 | 3, 4 | 344 | ||
GenBank mRNA | AK049183.1 | - | 3, 4 | 332 | ||
AK145787.1 | - | 3, 4 | 344 |
Gene Symbol: Zfp874b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076791.2 | NP_001070259.1 | 3, 4 | 341 | ||
GenBank mRNA | AK137581.1 | - | 3, 4 | 319 |
