siRNA Details
| Gene Symbol: Mb21d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177718.3 | NP_808386.2 | 2 | 679 | ||
| GenBank mRNA | AK079722.1 | - | 3 | 639 | ||
| BC068245.1 | - | 2 | 667 | |||
| BC085092.1 | - | 2 | 605 | 30735763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mb21d2| Gene Symbol: Mb21d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177718.3 | NP_808386.2 | 2 | 679 | ||
| GenBank mRNA | AK079722.1 | - | 3 | 639 | ||
| BC068245.1 | - | 2 | 667 | |||
| BC085092.1 | - | 2 | 605 | 30735763 | ||