siRNA Details
| Gene Symbol: Slc22a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039371.2 | NP_001034460.2 | 10, 11 | 1711 | ||
| GenBank mRNA | AK029695.1 | - | 7, 8 | 873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a15| Gene Symbol: Slc22a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039371.2 | NP_001034460.2 | 10, 11 | 1711 | ||
| GenBank mRNA | AK029695.1 | - | 7, 8 | 873 | ||