siRNA Details
| Gene Symbol: Frmpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177750.6 | NP_808418.1 | 5 | 571 | ||
| GenBank mRNA | AK047241.1 | - | 3 | 2291 | ||
| BC117734.1 | - | 3 | 227 | 40089821 | ||
| BC117735.1 | - | 3 | 227 | 40089822 | ||
| BC129957.1 | - | 3 | 227 | 40089827 | ||
| BC129958.1 | - | 3 | 227 | 40089829 | ||

Frmpd3
Genomic Map