siRNA Details
| Gene Symbol: Zfp385c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177790.4 | NP_808458.2 | 5, 6 | 940 | ||
| GenBank mRNA | AK044299.1 | - | 5, 6 | 941 | ||
| BC116726.1 | - | 5, 6 | 901 | 40106534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp385c| Gene Symbol: Zfp385c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177790.4 | NP_808458.2 | 5, 6 | 940 | ||
| GenBank mRNA | AK044299.1 | - | 5, 6 | 941 | ||
| BC116726.1 | - | 5, 6 | 901 | 40106534 | ||