siRNA Details
| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031395.1 | - | 10 | 1406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcmdc2| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031395.1 | - | 10 | 1406 | ||