siRNA Details
| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177730.3 | NP_808398.1 | 5 | 1693 | ||
| GenBank mRNA | AK039735.1 | - | 1 | 345 | ||
| AK041202.1 | - | 118 | ||||
| AK045511.1 | - | 5 | 1743 | |||
| BC048776.1 | - | 5 | 1693 | 6439769 | ||
| BC138209.1 | - | 5 | 1550 | 8861228 | ||
| BC145952.1 | - | 5 | 1550 | 40131255 | ||

Impad1
Genomic Map