siRNA Details
| Gene Symbol: Triml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177742.4 | NP_808410.1 | 2, 3 | 627 | ||
| GenBank mRNA | AK077092.1 | - | 2, 3 | 629 | ||
| BC050188.1 | - | 1 | 242 | |||
| BC141202.1 | - | 2, 3 | 606 | 9055814 | ||
| BC145531.1 | - | 3 | 609 | 9054094 | ||

Triml1
Genomic Map