siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177765.3 | NP_808433.2 | 10 | 1588 | ||
| GenBank mRNA | AK029565.1 | - | 10 | 1587 | ||
| AK161519.1 | - | 10 | 1411 | |||
| AM690757.1 | - | 9 | 1289 | |||
| BC145749.1 | - | 9 | 1196 | |||

Ttll13
Genomic Map