siRNA Details
| Gene Symbol: Bpifc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177772.4 | NP_808440.2 | 16 | 1754 | ||
| GenBank mRNA | AK028764.1 | - | 16 | 1751 | ||
| AK132518.1 | - | 16 | 1790 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifc| Gene Symbol: Bpifc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177772.4 | NP_808440.2 | 16 | 1754 | ||
| GenBank mRNA | AK028764.1 | - | 16 | 1751 | ||
| AK132518.1 | - | 16 | 1790 | |||