siRNA Details
| Gene Symbol: BC049635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177785.4 | NP_808453.1 | 3 | 255 | ||
| GenBank mRNA | AK077002.1 | - | 3 | 256 | ||
| BC049635.1 | - | 3 | 247 | 6774793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049635| Gene Symbol: BC049635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177785.4 | NP_808453.1 | 3 | 255 | ||
| GenBank mRNA | AK077002.1 | - | 3 | 256 | ||
| BC049635.1 | - | 3 | 247 | 6774793 | ||