siRNA Details
| Gene Symbol: Tmem26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177794.3 | NP_808462.2 | 6 | 2844 | ||
| GenBank mRNA | AK028283.1 | - | 6 | 2845 | ||
| AK084351.1 | - | 6 | 2822 | |||
| AK143866.1 | - | 6 | 2822 | |||
| AK156799.1 | - | 1 | 1615 | |||

Tmem26
Genomic Map