siRNA Details
| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037190.1 | - | 4 | 3099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5084| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037190.1 | - | 4 | 3099 | ||