siRNA Details
| Gene Symbol: Apol10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177820.4 | NP_808488.2 | 4 | 883 | ||
| GenBank mRNA | AK033678.1 | - | 855 | |||
| AK144113.1 | - | 4 | 887 | |||
| BC147348.1 | - | 829 | 9056388 | |||
| BC147349.1 | - | 829 | 9056408 | |||

Apol10b
Genomic Map