siRNA Details
| Gene Symbol: C130021I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081497.1 | - | 1789 | |||
| AK147796.1 | - | 3 | 2601 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130021I20Rik| Gene Symbol: C130021I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081497.1 | - | 1789 | |||
| AK147796.1 | - | 3 | 2601 | |||