siRNA Details
| Gene Symbol: 4933411G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177880.4 | NP_808548.1 | 3 | 777 | ||
| GenBank mRNA | AK030175.1 | - | 3 | 778 | ||
| BC115568.1 | - | 3 | 310 | 40102090 | ||
| BC115569.1 | - | 3 | 399 | 40102091 | ||

4933411G11Rik
Genomic Map