siRNA Details
Gene Symbol: A530021J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040731.1 | - | 2 | 251 | ||
BC145053.1 | - | 2 | 149 | |||
BC145836.1 | - | 2 | 149 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A530021J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040731.1 | - | 2 | 251 | ||
BC145053.1 | - | 2 | 149 | |||
BC145836.1 | - | 2 | 149 |