siRNA Details
| Gene Symbol: Dgkk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177914.3 | NP_808582.3 | 4959 | |||
| GenBank mRNA | AK081332.1 | - | 1 | 167 | ||
| BC075627.1 | - | 4959 | 6852516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkk| Gene Symbol: Dgkk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177914.3 | NP_808582.3 | 4959 | |||
| GenBank mRNA | AK081332.1 | - | 1 | 167 | ||
| BC075627.1 | - | 4959 | 6852516 | |||