siRNA Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177884.3 | NP_808552.2 | 8 | 1204 | ||
| GenBank mRNA | AK038369.1 | - | 9 | 1277 | ||
| AK050789.1 | - | 8 | 1146 | |||
| AK077263.1 | - | 8 | 1194 | |||
| AK078171.1 | - | 8 | 1195 | |||
| AK136387.1 | - | 6 | 806 | |||
| AK164830.1 | - | 1 | 1628 | |||

Gcfc2
Genomic Map