siRNA Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177887.4 | NP_808555.1 | 5 | 870 | ||
| GenBank mRNA | AK041846.1 | - | 5 | 871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem150b| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177887.4 | NP_808555.1 | 5 | 870 | ||
| GenBank mRNA | AK041846.1 | - | 5 | 871 | ||