siRNA Details
Gene Symbol: Gm14461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177843.3 | NP_808511.1 | 6 | 1759 | ||
GenBank mRNA | AK044135.1 | - | 6 | 1760 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm14461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177843.3 | NP_808511.1 | 6 | 1759 | ||
GenBank mRNA | AK044135.1 | - | 6 | 1760 |