siRNA Details
| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001415.3 | NP_001406.1 | 4 | 393 | ||
| GenBank mRNA | AB451230.1 | - | 4 | 372 | ||
| AB451353.1 | - | 4 | 372 | |||
| AK222577.1 | - | 4 | 385 | |||
| AK290397.1 | - | 4 | 385 | |||
| AK312247.1 | - | 4 | 385 | |||
| BC019906.1 | - | 4 | 393 | 4419438 | ||
| L19161.1 | - | 4 | 372 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK076900.

EIF2S3
Genomic Map