siRNA Details
| Gene Symbol: Gm9801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086712.1 | - | 2920 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9801| Gene Symbol: Gm9801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086712.1 | - | 2920 | |||