siRNA Details
| Gene Symbol: D830050J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052941.1 | - | 965 | |||
| AK085558.1 | - | 998 | ||||
| AK142706.1 | - | 964 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D830050J10Rik| Gene Symbol: D830050J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052941.1 | - | 965 | |||
| AK085558.1 | - | 998 | ||||
| AK142706.1 | - | 964 | ||||