siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178080.4 | NP_835181.2 | 1 | 682 | ||
| GenBank mRNA | AK028810.1 | - | 1 | 650 | ||
| AK042081.1 | - | 1 | 658 | |||
| AK122286.1 | - | 1 | 677 | |||
| BC025653.1 | - | 1 | 608 | |||
| BC050783.1 | - | 1 | 636 | |||
| BC057350.1 | - | 1 | 599 | |||
| BC141172.1 | - | 1 | 610 | |||

Pde4dip
Genomic Map