siRNA Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001444.2 | NP_001435.1 | 4 | 553 | ||
GenBank mRNA | AK226036.1 | - | 4 | 502 | ||
BC019385.2 | - | 4 | 486 | 3862100 | ||
BC070303.1 | - | 1 | 500 | 6496304 | ||
M94856.1 | - | 4 | 488 |
