siRNA Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122730.2 | NP_001116202.2 | 4 | 1553 | ||
| NM_178242.2 | NP_839973.1 | 4 | 1553 | |||
| GenBank mRNA | AK141849.1 | - | 1 | 644 | ||
| BC049818.1 | - | 4 | 1553 | 6511384 | ||

Tnrc18
Genomic Map