siRNA Details
| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ554212.1 | - | 1 | 191 | ||
| BC141344.1 | - | 1 | 240 | 9055956 | ||
| BC141345.1 | - | 1 | 241 | 9055957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Teddm1a| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ554212.1 | - | 1 | 191 | ||
| BC141344.1 | - | 1 | 240 | 9055956 | ||
| BC141345.1 | - | 1 | 241 | 9055957 | ||