siRNA Details
| Gene Symbol: FABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319039.1 | NP_001305968.1 | 2 | 424 | ||
| NM_001319041.1 | NP_001305970.1 | 2 | 424 | |||
| NM_001319042.1 | NP_001305971.1 | 2 | 165 | |||
| NM_001446.4 | NP_001437.1 | 2 | 424 | |||
| GenBank mRNA | AB208815.1 | - | 2 | 171 | ||
| AI879148.1 | - | 2 | 467 | |||
| AJ002962.1 | - | 2 | 180 | |||
| AK289836.1 | - | 2 | 183 | |||
| AK307272.1 | - | 2 | 184 | |||
| AK311867.1 | - | 2 | 183 | |||
| AL512688.1 | - | 2 | 140 | |||
| BC012299.1 | - | 2 | 290 | 4707233 | ||
| BI860090.1 | - | 2 | 398 | |||
| BX461253.2 | - | 2 | 145 | |||
| CR457057.1 | - | 2 | 104 | |||
| D50373.1 | - | 2 | 156 | |||
| D88648.1 | - | 2 | 155 | |||
| KM111186.1 | - | 2 | 165 | |||
| U51338.1 | - | 2 | 159 | |||
| U81235.1 | - | 2 | 119 | |||

FABP7
Genomic Map