siRNA Details
| Gene Symbol: Zfp74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178384.3 | NP_848471.2 | 5 | 791 | ||
| GenBank mRNA | BC050048.1 | - | 5 | 791 | 30011303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp74| Gene Symbol: Zfp74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178384.3 | NP_848471.2 | 5 | 791 | ||
| GenBank mRNA | BC050048.1 | - | 5 | 791 | 30011303 | |