siRNA Details
| Gene Symbol: Tmem202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178388.2 | NP_848475.1 | 3 | 517 | ||
| GenBank mRNA | AK008510.1 | - | 392 | |||
| AK015202.1 | - | 3 | 471 | |||
| BC049695.1 | - | 3 | 517 | 6705996 | ||
| BC132482.1 | - | 3 | 504 | 40130759 | ||
| BC138196.1 | - | 3 | 504 | 8861215 | ||

Tmem202
Genomic Map