siRNA Details
Gene Symbol: Ccdc144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178418.4 | NP_848505.2 | 5 | 652 | ||
GenBank mRNA | AK133178.1 | - | 5 | 638 | ||
BC050811.1 | - | 5 | 654 | 6772771 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178418.4 | NP_848505.2 | 5 | 652 | ||
GenBank mRNA | AK133178.1 | - | 5 | 638 | ||
BC050811.1 | - | 5 | 654 | 6772771 |