siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178599.4 | NP_848714.1 | 2, 3 | 263 | ||
| GenBank mRNA | AK009275.1 | - | 2, 3 | 252 | ||
| AK012653.1 | - | 2, 3 | 249 | |||
| AK013371.1 | - | 2, 3 | 236 | |||
| AK077934.1 | - | 2, 3 | 263 | |||
| AK151560.1 | - | 2, 3 | 246 | |||
| AK170011.1 | - | 2, 3 | 236 | |||
| BC098183.1 | - | 2, 3 | 239 | 5150466 | ||

Commd8
Genomic Map