siRNA Details
| Gene Symbol: Mrpl50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178603.4 | NP_848718.1 | 2 | 1818 | ||
| GenBank mRNA | AK002646.1 | - | 2 | 1806 | ||
| AK007405.1 | - | 2 | 1815 | |||
| AK031229.1 | - | 2 | 1819 | |||
| AK049421.1 | - | 2 | 1808 | |||
| BC021318.1 | - | 2 | 1825 | 5035369 | ||

Mrpl50
Genomic Map