siRNA Details
| Gene Symbol: Tcfl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178254.3 | NP_839985.2 | 5, 6 | 1457 | ||
| GenBank mRNA | AY234363.1 | - | 5, 6 | 1251 | ||
| BC108399.1 | - | 5, 6 | 1457 | 6394526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcfl5| Gene Symbol: Tcfl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178254.3 | NP_839985.2 | 5, 6 | 1457 | ||
| GenBank mRNA | AY234363.1 | - | 5, 6 | 1251 | ||
| BC108399.1 | - | 5, 6 | 1457 | 6394526 | ||