siRNA Details
| Gene Symbol: Flrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172160.1 | NP_001165631.1 | 3 | 3610 | ||
| NM_178382.4 | NP_848469.1 | 3 | 3057 | |||
| GenBank mRNA | AK017456.1 | - | 1 | 408 | ||
| AK028252.1 | - | 3 | 3051 | |||
| AK031464.1 | - | 3 | 3051 | |||
| AK161100.1 | - | 3 | 3053 | |||

Flrt3
Genomic Map