siRNA Details
| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178398.4 | NP_848485.1 | 3 | 429 | ||
| GenBank mRNA | AK010881.1 | - | 3 | 457 | ||
| AK043857.1 | - | 3 | 462 | |||
| AK155113.1 | - | 3 | 459 | |||
| AK169051.1 | - | 3 | 403 | |||
| AK172030.1 | - | 3 | 405 | |||
| AK194559.1 | - | 2 | 158 | |||
| BC044894.1 | - | 3 | 454 | 4235237 | ||

Wipi2
Genomic Map