siRNA Details
| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040663.2 | NP_001035753.1 | 5 | 468 | ||
| NR_102272.1 | 6 | 611 | ||||
| GenBank mRNA | BC036094.1 | - | 6 | 599 | 5296612 | |
| BC069470.1 | - | 5 | 419 | 7262175 | ||
| U19142.1 | - | 6 | 533 | |||

GAGE1
Genomic Map